Learn More
SEKISUI Diagnostics Metrix™ Reader
Catalog No. 22321139 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Sample Type:
Anterior Nasal Swab
Catalog No. Sample Type
22-321-139 Anterior Nasal Swab
1 options
This item is not returnable. View return policy
Catalog No. 22-321-139 Supplier SEKISUI Diagnostics Supplier No. MTRXRDR
This item is not returnable. View return policy
The Metrix™ reader is compact and robust and ideal for professional use in diverse locations, including clinics and mobile health units. It’s a maintenance free device with no calibration step required.
  • Portable - A compact device that is powered by a USB-C power cord.
  • Eco-friendly - A reusable device that does not have to be thrown away after each use; no batteries required.
  • Intuitive - A maintenance free device with no calibration step required.
  • Convenient - Multicolor test status indicators with direct printed labels.
TRUSTED_SUSTAINABILITY
Specifications
Type Accuracy Reader
Sample Type Anterior Nasal Swab