Learn More
Cayman Chemical MetrIzamIde 10mg
SDP
Catalog No. 502939443
Encompass_Preferred
A non-ionic contrast agent; does not induce EEG abnormalities in rabbits at 0.1 ml/kg of a 370 mg I/ml solution; has been used as a solute in gradient centrifugation applications
Catalog No. 50-293-9443 Supplier Cayman Chemical Supplier No. 2772410 MG
A non-ionic contrast agent; does not induce EEG abnormalities in rabbits at 0.1 ml/kg of a 370 mg I/ml solution; has been used as a solute in gradient centrifugation applications