Learn More
Medchemexpress LLC HY-B1699 100mg Medchemexpress, Metrizoic acid CAS:1949-45-7 Purity:>98%
SDP
Catalog No. 501970157
Encompass_Preferred
Medchemexpress, HY-B1699 100mg Metrizoic acid CAS:1949-45-7 Metrizoic acid (Metrizoate) is an ionic contrast medium. Metrizoic acid (Metrizoate) shows high osmolality and has a risk of inducing allergic reactions[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0157 Supplier Medchemexpress LLC Supplier No. HYB1699100MG
Medchemexpress, HY-B1699 100mg Metrizoic acid CAS:1949-45-7 Metrizoic acid (Metrizoate) is an ionic contrast medium. Metrizoic acid (Metrizoate) shows high osmolality and has a risk of inducing allergic reactions[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.