Learn More
Medchemexpress LLC Metrizoic acid | 1949-45-7 | MFCD00867964 | 98.0% | 250 MG
SDP
Catalog No. 5000414229
Encompass_Preferred
Metrizoic acid (Metrizoate) is an ionic contrast medium that exhibits high osmolality and carries a risk of inducing allergic reactions. It is supplied as a solid, white to off-white powder with a purity of 98.0% and is stable under recommended storage conditions.
  • High osmolality ionic contrast medium.
  • Risk of inducing allergic reactions.
  • Solid, white to off-white appearance.
  • Purity of 98.0%.
  • Stable under recommended storage conditions.
Catalog No. 50-004-14229 Supplier Medchemexpress LLC Supplier No. 250MGHYB1699
Metrizoic acid (Metrizoate) is an ionic contrast medium that exhibits high osmolality and carries a risk of inducing allergic reactions. It is supplied as a solid, white to off-white powder with a purity of 98.0% and is stable under recommended storage conditions.
  • High osmolality ionic contrast medium.
  • Risk of inducing allergic reactions.
  • Solid, white to off-white appearance.
  • Purity of 98.0%.
  • Stable under recommended storage conditions.