Learn More
MSC Metro Adjustable Chrome Wire Shelf, 18x48 in, 800 lb Capacity
Catalog No. 5000529915
Encompass
This Metro Super Adjustable Chrome Wire Shelf offers a rigid, innovative wire storage system with a unique corner release system, enabling tool-free shelf adjustments. Measuring 18 x 48 in, this shelf supports up to 800 lb and is ideal for dry storage environments. It is designed to work with the entire Super Erecta system of shelves and accessories and is NSF Certified.
  • Tool-free adjustment with a unique corner release system.
  • Durable heavy-duty wire construction.
  • Sleek chrome finish.
  • 800 lb weight capacity.
  • Compatible with the entire Super Erecta system.
  • Includes split sleeves for secure fit.
  • Ideal for dry storage environments.
  • NSF certified.
Catalog No. 50-005-29915 Supplier MSC Supplier No. 03030558
This Metro Super Adjustable Chrome Wire Shelf offers a rigid, innovative wire storage system with a unique corner release system, enabling tool-free shelf adjustments. Measuring 18 x 48 in, this shelf supports up to 800 lb and is ideal for dry storage environments. It is designed to work with the entire Super Erecta system of shelves and accessories and is NSF Certified.
  • Tool-free adjustment with a unique corner release system.
  • Durable heavy-duty wire construction.
  • Sleek chrome finish.
  • 800 lb weight capacity.
  • Compatible with the entire Super Erecta system.
  • Includes split sleeves for secure fit.
  • Ideal for dry storage environments.
  • NSF certified.