Learn More
Metro™ Double-Deep qwikTRAK™ Stationary End Unit Kits
Catalog No. 17823734 Shop All Metro Products
Change view
Click to view available options
Material:
Chrome
Polymer
Stainless Steel
Steel
For Use With (Equipment):
Metro MetroMax i and Q Shelving System
Metro Super Erecta, Super Adjustable Shelving System
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Equipment)
17-823-734 Stainless Steel Metro Super Erecta, Super Adjustable Shelving System
17-823-732 Chrome Metro Super Erecta, Super Adjustable Shelving System
17-823-733 Steel Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
17-823-736 Polymer Metro MetroMax i and Q Shelving System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-823-734 Supplier Metro™ Supplier No. LBTES
Add to Cart
Add to Cart
Connects end units to tracks and anchors tracks to floor
  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
  • Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Product Type Stationary End Unit Kit
For Use With (Equipment) Metro Super Erecta, Super Adjustable Shelving System
Includes 74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary end units to track and anchors track to floor
Weight (English) 73.6 lb.
Weight (Metric) 33.5 kg
Material Stainless Steel