Learn More
Metro™ Double-Deep qwikTRAK™ Stationary Intermediate Unit Kits
GSA_VA
Catalog No. 17823737 Shop All Metro Products
Change view
Click to view available options
Finish:
Chrome-plated
Heavy Polymer
Metroseal Green Epoxy
Stainless Steel
For Use With (Equipment):
Metro MetroMax i and Q Shelving System
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Finish For Use With (Equipment)
17-823-737 Chrome-plated Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
17-823-738 Metroseal Green Epoxy Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
17-823-739 Stainless Steel Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
17-823-741 Heavy Polymer Metro MetroMax i and Q Shelving System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-823-737 Supplier Metro™ Supplier No. LBTAC
Add to Cart
Add to Cart
Connects one double-deep stationary imtermediate unit to to tracks and anchors tracks to floor
  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ and MetroMax iQ shelving systems
  • Kit includes 74 in. (188cm) high stationary posts and connecting hardware
Order Info
Shelves are sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Stationary Intermediate Unit Kit
Finish Chrome-plated
For Use With (Equipment) Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
Includes 74 in. High stationary posts and connecting hardware to connect double-deep qwikTRAK™ stationary intermediate units to track and anchors track to floor
Weight (English) 39.6 lb.
Weight (Metric) 18 kg
Material Steel