Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ Flexline™ Sharps Container Cart Accessory

Catalog No. 13451217 Shop All Metro Products
Change view
Click to view available options
Includes:
Sharps Container Bracket with Hook-and-Loop Straps
Sharps Container Only
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
13-451-217 Sharps Container Bracket with Hook-and-Loop Straps
13-451-216 Sharps Container Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-451-217 Supplier Metro™ Supplier No. FL250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sharps accessories attach to carts for easy accessibility while mobile.

  • Sharps cart accessory in lockable or non-locking styles
  • Lockable dimensions with glove box: 12.06 x 33.02 x 50.48 cm (4.75 x 13 x 19.875 in.)
  • Lockable dimensions without glove box: 12.06 x 33.02 x 28.89 cm (4.75 x 13 x 11.375 in.)
  • Nonlocking dimensions: 24.1 x 8.3 x 23.7cm (9.5 x 3.25 9.3125 in.)

Specifications

Color Black
Autoclavable Not Autoclavable
Includes Sharps Container Bracket with Hook-and-Loop Straps
For Use With (Equipment) Lifeline Carts
Material Epoxy-coated Steel Wire
Product Type Sharps Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.