Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax i Basket Glassware Cart
GSA_VA

Catalog No. 13451093 Shop All Metro Products
Change view
Click to view available options
Height (English):
40 in.
60 in.
Description:
2-Basket Unit
2-Tote Kit Unit
4-Basket Unit
4-Tote Kit Unit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Description
13-451-093 40 in. 2-Tote Kit Unit
13-451-092 60 in. 4-Basket Unit
13-451-091 40 in. 2-Basket Unit
13-451-094 60 in. 4-Tote Kit Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-451-093 Supplier Metro™ Supplier No. GWTX31
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Level 2 cart with built-in Microban antimicrobial product protection allows arrangement configuration, ensuring storage and movement won't disrupt your workday.

  • Modular design offers ultimate flexibility.
  • Reinforced polymers provide superior strength (up to 800 lb. per shelf).
  • Corrosion-proof, suitable for damp and humid conditions.
  • Lifetime warranty against rust and corrosion.
  • Shelves wipe down easily and have a retaining edge that contains drips and spills.
  • Two-basket and four-basket capacity glassware solutions are offered in a spacesaving 18 x 36 in. (457 x 914mm) footprint.
  • Solid MetroMax bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning.
  • Wide range of accessories available separately, including all-polymer casters for maximum corrosion resistance and premium performance.

Order Info

Casters not included (5PC Series Metro casters are recommended for superior lifetime performance).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Glassware Cart
Capacity (Metric) 362.8 kg
Color Taupe, Blue
Description 2-Tote Kit Unit
Dimensions (L x W x H) 36 x 18 x 4 in. (91.4 x 45.7 x 10.2 cm)
For Use With (Application) Glassware Transportation
Height (English) 40 in.
Height (Metric) 101.6 cm
Includes 3 Solid MetroMax i™ bottom shelves, 2 tote glassware transport kits (large and small tote with interior dividers), 4 polymer casters (2-swivel/2-swivel brake), all-polymer horn, stainless steel axle
Length (English) 36 in.
Length (Metric) 91.4 cm
Load Bearing Capacity (English) 800 lb.
Load Bearing Capacity (Metric) 362.8 kg
Material Polymer
No. of Shelves 3
Width (English) 18 in.
Width (Metric) 45.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.