Learn More
Metro™ MetroMax i™ Top-Track Mobile Unit Kits
GSA_VA
Catalog No. 17820340 Shop All Metro Products
Change view
Click to view available options
Width (English) Shelf:
18 in.
24 in.
Includes:
Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Shelf Includes
17-820-340 24 in. Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
17-820-337 18 in. Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
17-820-338 18 in. Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
17-820-339 24 in. Posts, Chrome-plated rigid casters, caster channel, bumpers and roller assemblies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-820-340 Supplier Metro™ Supplier No. MXTTM24S
Add to Cart
Add to Cart
Includes
Posts, rigid casters, caster channels, bumpers and roller assemblies
High-density, easy access storage
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
Order Info
  • Order one per mobile unit
  • Shelves sold separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Mobile Unit Kit
For Use With (Equipment) Metro™ MetroMax i™ and Q™ Top-Track™ system
Includes Posts, Stainless Steel rigid casters, caster channel, bumpers and roller assemblies
Width (English) Shelf 24 in.
Width (Metric) Shelf 61 cm