Learn More
MSC Metro MetroMax Q Post, Epoxy-Coated Steel, 15-Year Warranty, 1 inch Adjustment
Catalog No. 5000512179
Encompass
MetroMax Q Posts are key components of the versatile MetroMax Q Shelving System, designed to provide durable and corrosion-resistant storage solutions. These posts allow for easy and flexible shelf configuration with 1-inch adjustments, making them suitable for various demanding environments.
  • Constructed from epoxy-coated steel.
  • Offers a 15-year warranty against rust and corrosion for epoxy-coated steel posts.
  • Optional polymer posts provide a lifetime warranty against corrosion.
  • Shelves are adjustable in 1-inch increments along the posts.
  • Available in stationary and mobile configurations.
  • Ideal for wet or humid environments, walk-in coolers, and sterile areas.
  • Not compatible with old style MetroMax systems.
Catalog No. 50-005-12179 Supplier MSC Supplier No. 66688664
MetroMax Q Posts are key components of the versatile MetroMax Q Shelving System, designed to provide durable and corrosion-resistant storage solutions. These posts allow for easy and flexible shelf configuration with 1-inch adjustments, making them suitable for various demanding environments.
  • Constructed from epoxy-coated steel.
  • Offers a 15-year warranty against rust and corrosion for epoxy-coated steel posts.
  • Optional polymer posts provide a lifetime warranty against corrosion.
  • Shelves are adjustable in 1-inch increments along the posts.
  • Available in stationary and mobile configurations.
  • Ideal for wet or humid environments, walk-in coolers, and sterile areas.
  • Not compatible with old style MetroMax systems.