Learn More
Metrohm USA Metrosep A Supp 17 - 150/4.0, 1 each
SDP
Supplier:  Metrohm USA 601032420
Encompass
The Metrosep A Supp 17 - 150/4.0 separation column is the column of choice for anion determinations that require good separating efficiency and short separation times at room temperature. The maximum flow rate of 1.4 mL/min then also makes it possible to optimize the determination. The Metrosep A-Supp-17 columns convince with their good price-performance ratio.
Catalog No. NC2019731