Learn More
MSC Metro Shelf, MetroMax i Polymer, Grid Mat, 18 in x 72 in
Catalog No. 5000527412
Encompass
This shelf is a component of the MetroMax i Polymer Shelving system, recognized as a strong, versatile, and corrosion-proof solution. It is constructed from advanced polymers and Type 304 stainless corners, and features a grid mat. The shelf mats include built-in Microban antimicrobial product protection, and this 72-inch shelf has a maximum weight capacity of 750 lb (340 kg).
  • Adjusts in 1 inch increments
  • Removable, easy-to-clean open grid mats promote air circulation and light penetration
  • Lifetime warranty against rust and corrosion
  • Includes removable mats and one bag of wedges
  • Built-in antimicrobial product protection
  • Constructed from advanced polymers and Type 304 stainless corners
Catalog No. 50-005-27412 Supplier MSC Supplier No. 66688540
This shelf is a component of the MetroMax i Polymer Shelving system, recognized as a strong, versatile, and corrosion-proof solution. It is constructed from advanced polymers and Type 304 stainless corners, and features a grid mat. The shelf mats include built-in Microban antimicrobial product protection, and this 72-inch shelf has a maximum weight capacity of 750 lb (340 kg).
  • Adjusts in 1 inch increments
  • Removable, easy-to-clean open grid mats promote air circulation and light penetration
  • Lifetime warranty against rust and corrosion
  • Includes removable mats and one bag of wedges
  • Built-in antimicrobial product protection
  • Constructed from advanced polymers and Type 304 stainless corners