Learn More
MSC Metro Shelf, Polymer, Removable Grid Mat, 24 x 60 in
Catalog No. 5000510239
Encompass
The MetroMax Q shelf is an extra shelf designed for use with MetroMax Q shelving systems. It features polymer construction and removable grid mats, offering corrosion resistance and Microban antimicrobial protection.
  • Removable, wash-machine friendly mats.
  • Shelves adjust at 1 inch increments.
  • Built-in antimicrobial product protection helps inhibit the growth of odor and stain-causing bacteria, mold, mildew, and fungi.
  • Corrosion-resistant polymer and coated wire with removable shelf mats.
  • Compatible with other shelving systems.
  • Supports HACCP processes.
  • Interchangeable shelves and posts offer increased storage efficiencies.
  • Includes a 15-year corrosion warranty.
Catalog No. 50-005-10239 Supplier MSC Supplier No. 66688458
The MetroMax Q shelf is an extra shelf designed for use with MetroMax Q shelving systems. It features polymer construction and removable grid mats, offering corrosion resistance and Microban antimicrobial protection.
  • Removable, wash-machine friendly mats.
  • Shelves adjust at 1 inch increments.
  • Built-in antimicrobial product protection helps inhibit the growth of odor and stain-causing bacteria, mold, mildew, and fungi.
  • Corrosion-resistant polymer and coated wire with removable shelf mats.
  • Compatible with other shelving systems.
  • Supports HACCP processes.
  • Interchangeable shelves and posts offer increased storage efficiencies.
  • Includes a 15-year corrosion warranty.