Learn More
MSC Metro Super Erecta Wire Shelf, Steel, Chrome, 18 x 60 in
Catalog No. 5000529438
Encompass
This Metro Super Erecta Wire Shelf is constructed from steel with a chrome finish, designed for use with Intermetro Shelving systems. It measures 18 inches deep by 60 inches wide.
  • Constructed from durable steel.
  • Features a chrome finish for enhanced durability.
  • Designed for use with Intermetro Shelving systems.
  • Measures 18 inches deep by 60 inches wide.
Catalog No. 50-005-29438 Supplier MSC Supplier No. 03030186
This Metro Super Erecta Wire Shelf is constructed from steel with a chrome finish, designed for use with Intermetro Shelving systems. It measures 18 inches deep by 60 inches wide.
  • Constructed from durable steel.
  • Features a chrome finish for enhanced durability.
  • Designed for use with Intermetro Shelving systems.
  • Measures 18 inches deep by 60 inches wide.