Learn More
MSC Metro Super Erecta Wire Shelf, Steel, Zinc-Plated, 24 x 36 in
Catalog No. 5000533466
Encompass
This wire shelf is designed for use with Intermetro shelving systems, providing an efficient and organized storage solution. Constructed from durable steel with a zinc-plated finish, it offers reliable performance for various applications. Its open-wire design facilitates air circulation and visibility of stored items.
  • Open-wire design minimizes dust and allows for greater visibility of contents.
  • Constructed from durable steel for long-lasting use.
  • Features a protective zinc-plated finish.
  • Measures 24 inches in depth and 36 inches in width.
  • Has a height of 1-1/2 inches.
  • Compatible with Intermetro shelving units.
Catalog No. 50-005-33466 Supplier MSC Supplier No. 03030061
This wire shelf is designed for use with Intermetro shelving systems, providing an efficient and organized storage solution. Constructed from durable steel with a zinc-plated finish, it offers reliable performance for various applications. Its open-wire design facilitates air circulation and visibility of stored items.
  • Open-wire design minimizes dust and allows for greater visibility of contents.
  • Constructed from durable steel for long-lasting use.
  • Features a protective zinc-plated finish.
  • Measures 24 inches in depth and 36 inches in width.
  • Has a height of 1-1/2 inches.
  • Compatible with Intermetro shelving units.