Learn More
MSC MetroVac Microfiber Vacuum Filter Bag, Disposable, Microfiber, 5 Bags
Catalog No. 502818593
Encompass
This microfiber vacuum filter bag is designed for effective filtration in various cleaning applications. Constructed from fine synthetic fibers, it offers high absorbency and is suitable for heavier cleaning tasks.
  • Disposable filter bag
  • Made from microfiber material
  • Highly absorbent, suitable for heavier cleaning, disinfecting, and scrubbing applications
  • Compatible with DV3ESD-1, MDV-3, and MDV-2 vacuum models
Catalog No. 50-281-8593 Supplier MSC Supplier No. 00503227
This microfiber vacuum filter bag is designed for effective filtration in various cleaning applications. Constructed from fine synthetic fibers, it offers high absorbency and is suitable for heavier cleaning tasks.
  • Disposable filter bag
  • Made from microfiber material
  • Highly absorbent, suitable for heavier cleaning, disinfecting, and scrubbing applications
  • Compatible with DV3ESD-1, MDV-3, and MDV-2 vacuum models