Learn More
MSC Metro Wire Shelf, 18 x 54 inch, Steel, Brite Finish
Catalog No. 5000529640
Encompass
This wire shelf features an open wire design that minimizes dust accumulation and allows free air circulation, enhancing visibility and light penetration for stored items. Constructed from steel with a durable brite finish, it is designed for use with shelving systems.
  • Minimizes dust accumulation and allows free air circulation
  • Enhances visibility of stored items
  • Maximizes light penetration
  • Can be reconfigured with various accessories
  • Assembles quickly without special tools
  • Features shelf ribs that run front to back for smooth item movement
  • Allows loading and unloading from all sides, maximizing storage space
  • Shelves are adjustable at 1-inch intervals along the post length
  • Includes bolt levelers to compensate for surface irregularities
  • Meets U.S. government specifications MIL-S-40144E
  • Equipped with a clear protective coating
Catalog No. 50-005-29640 Supplier MSC Supplier No. 03030012
This wire shelf features an open wire design that minimizes dust accumulation and allows free air circulation, enhancing visibility and light penetration for stored items. Constructed from steel with a durable brite finish, it is designed for use with shelving systems.
  • Minimizes dust accumulation and allows free air circulation
  • Enhances visibility of stored items
  • Maximizes light penetration
  • Can be reconfigured with various accessories
  • Assembles quickly without special tools
  • Features shelf ribs that run front to back for smooth item movement
  • Allows loading and unloading from all sides, maximizing storage space
  • Shelves are adjustable at 1-inch intervals along the post length
  • Includes bolt levelers to compensate for surface irregularities
  • Meets U.S. government specifications MIL-S-40144E
  • Equipped with a clear protective coating