Learn More
Metrohm USA METROHM CAUTION GUARD COLUMN
SDP
Catalog No. NC0113983 Shop All Metrohm USA Products
Encompass
The Metrosep C 4 Guard/4.0 contains the C-4 column material and is used to protect all Metrosep cation columns that have a substrate based on silica gel. Particles and contaminations are reliably retained, allowing the lifetime of the analytical separation column to be prolonged considerably. The economical price is an additional plus. The Metrosep C 4 Guard/4.0 also functions according to the "On Column Guard System" and is screwed directly onto the separation column with nearly no dead volume.
Catalog No. NC0113983 Supplier Metrohm USA Supplier No. 61050500
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Metrosep C 4 Guard/4.0 contains the C-4 column material and is used to protect all Metrosep cation columns that have a substrate based on silica gel. Particles and contaminations are reliably retained, allowing the lifetime of the analytical separation column to be prolonged considerably. The economical price is an additional plus. The Metrosep C 4 Guard/4.0 also functions according to the "On Column Guard System" and is screwed directly onto the separation column with nearly no dead volume.