Learn More
Metrohm USA METROHM USA TX-900MH PRINTER
SDP
Supplier:  Metrohm USA 2.143.0200
Encompass
  • Compact impact printer for laboratory integration.
  • USB and RS232 interfaces for versatile connectivity.
  • Wired connectivity type.
  • Compatible with Metrohm 900 Touch Control, 915 KF Ti-Touch, 916 Ti-Touch.
  • Compatible with Metrohm 917 and 899 Coulometers with adapter.
  • Compatible with Metrohm 91X Meter, Eco Dosimat/Titrator, 949 pH Meter.
  • Includes RS-232 DB9 cable (6.2134.040) for connection.
  • Includes USB A-B 1.8 m cable (6.2151.020).
  • Manufacturer: Metrohm AG.
Catalog No. NC2885124