Learn More
MSC MetroMaxQ Shelf, Polymer, 18 inch Depth, 72 inch Width
Catalog No. 5000537440
Encompass
This extra shelf is designed for use with Metro Max Q shelving units. Constructed from polymer, it offers durability and features built-in antimicrobial protection, quick adjustment capabilities, and a removable design for ease of use and maintenance.
  • Microban antimicrobial protection
  • Quick adjustment for versatile configuration
  • Removable polymer shelf for easy cleaning and maintenance
  • Engineered for strength and durability
Catalog No. 50-005-37440 Supplier MSC Supplier No. 66688391
This extra shelf is designed for use with Metro Max Q shelving units. Constructed from polymer, it offers durability and features built-in antimicrobial protection, quick adjustment capabilities, and a removable design for ease of use and maintenance.
  • Microban antimicrobial protection
  • Quick adjustment for versatile configuration
  • Removable polymer shelf for easy cleaning and maintenance
  • Engineered for strength and durability