Learn More
Alkali Scientific Metronidazole, 100 G
SDP
Supplier:  Alkali Scientific MET001CP
Encompass_Preferred
Metronidazole, available in a 100g format, is an antibiotic commonly used to target anaerobic bacteria and certain protozoa. It works by disrupting the DNA of microorganisms, leading to cell death. Metronidazole is particularly useful in the treatment and research of infections caused by anaerobic bacteria, including those related to gastrointestinal, respiratory, and gynecological conditions. This compound is widely used in antimicrobial research to study its effects on bacterial and protozoal DNA synthesis and its role in resistance mechanisms. It is also used in clinical laboratories for susceptibility testing and in the study of microbial pathogenesis.
Catalog No. 50-006-59994