Learn More
Research Products International Corp Metronidazole, 100 Grams
SDP
Catalog No. 50213513
Encompass_Preferred
Metronidazole is a commonly used product in the life science marketplace with a broad range of applications.
It belongs to the class of drugs known as nitroimidazoles and exhibits potent antimicrobial activity against various anaerobic bacteria and parasites.
In the field of infectious diseases, metronidazole is primarily utilized to treat infections caused by anaerobic bacteria, such as Clostridium difficile, Bacteroides fragilis, and Helicobacter pylori. 
WARNING! This product can expose you to Metronidazole, a chemical which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-513 Supplier Research Products International Corp Supplier No. M81000100.0
Metronidazole is a commonly used product in the life science marketplace with a broad range of applications.
It belongs to the class of drugs known as nitroimidazoles and exhibits potent antimicrobial activity against various anaerobic bacteria and parasites.
In the field of infectious diseases, metronidazole is primarily utilized to treat infections caused by anaerobic bacteria, such as Clostridium difficile, Bacteroides fragilis, and Helicobacter pylori. 
WARNING! This product can expose you to Metronidazole, a chemical which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.