Learn More
Medchemexpress LLC HY-B0318 10g , Metronidazole CAS:443-48-1 Purity:>98%
SDP
Catalog No. 502259279
Encompass_Preferred
HY-B0318 10g Metronidazole CAS: 443-48-1 Metronidazole is an orally active nitroimidazole antibiotic, can be used to research anaerobic infections. Metronidazole can cross blood brain barrier and results inflammation and skeletal muscle contraction under long-term application[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9279 Supplier Medchemexpress LLC Supplier No. HYB031810G
HY-B0318 10g Metronidazole CAS: 443-48-1 Metronidazole is an orally active nitroimidazole antibiotic, can be used to research anaerobic infections. Metronidazole can cross blood brain barrier and results inflammation and skeletal muscle contraction under long-term application[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.