Learn More
Med Vet International Ayradia (Metronidazole) Oral Suspension, 125 mg/mL, 100 mL, Each
Catalog No. 502828790
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
AYRADIA™ (Metronidazole) Oral Suspension is a nitroimidazole for dogs, in the drug class anti-infectives, with antibacterial and antiprotozoal activities. The product is a flavored, oily suspension with brown visible particles Indicated for the treatment of Giardia duodenalis infection in dogs Shake vigorously before use Administered orally at a dose of 25 mg/kg (11.3 mg/lb) of body weight, using the supplied syringe
Catalog No. 50-282-8790 Supplier Med Vet International Supplier No. RXAYRADIA100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
AYRADIA™ (Metronidazole) Oral Suspension is a nitroimidazole for dogs, in the drug class anti-infectives, with antibacterial and antiprotozoal activities. The product is a flavored, oily suspension with brown visible particles Indicated for the treatment of Giardia duodenalis infection in dogs Shake vigorously before use Administered orally at a dose of 25 mg/kg (11.3 mg/lb) of body weight, using the supplied syringe