Learn More
Medchemexpress LLC Metronidazole | 443-48-1 | 50 G
SDP
Catalog No. 5000406341
Encompass_Preferred
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for the research of anaerobic infections.
  • Orally active nitroimidazole antibiotic
  • Crosses the blood-brain barrier
  • Used for research of anaerobic infections
  • Inhibits anaerobic protozoa (Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, Balantidium coli)
  • Inhibits anaerobic bacteria and demonstrates good bactericidal activity (4-8 μg/mL)
Catalog No. 50-004-06341 Supplier Medchemexpress LLC Supplier No. HYB031850G
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for the research of anaerobic infections.
  • Orally active nitroimidazole antibiotic
  • Crosses the blood-brain barrier
  • Used for research of anaerobic infections
  • Inhibits anaerobic protozoa (Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, Balantidium coli)
  • Inhibits anaerobic bacteria and demonstrates good bactericidal activity (4-8 μg/mL)