Learn More
Medchemexpress LLC Metronidazole | 443-48-1 | 99.96% | 100 G
SDP
Catalog No. 5000405826
Encompass_Preferred
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for research on anaerobic infections.
  • Exhibits inhibitory activity against anaerobic protozoa such as *Trichomonas vaginalis*, *Entamoeba histolytica*, *Giardia lamblia*, and *Balantidium coli*.
  • Inhibits anaerobic bacteria and shows good bactericidal activity (4-8 μg/mL).
  • Induces granular formation and triggers apoptosis in *Blastocystis sp.*.
Catalog No. 50-004-05826 Supplier Medchemexpress LLC Supplier No. HYB0318100G
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for research on anaerobic infections.
  • Exhibits inhibitory activity against anaerobic protozoa such as *Trichomonas vaginalis*, *Entamoeba histolytica*, *Giardia lamblia*, and *Balantidium coli*.
  • Inhibits anaerobic bacteria and shows good bactericidal activity (4-8 μg/mL).
  • Induces granular formation and triggers apoptosis in *Blastocystis sp.*.