Learn More
Medchemexpress LLC Metronidazole | 443-48-1 | 99.96% | 25 MG
SDP
Catalog No. 5000450589
Encompass_Preferred
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for research on anaerobic infections.
  • Inhibits anaerobic protozoa such as Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, and Balantidium coli
  • Inhibits anaerobic bacteria and demonstrates good bactericidal activity
  • Induces granular formation and triggers apoptosis in Blastocystis sp
  • Crosses the blood-brain barrier for neurotoxicity research
Catalog No. 50-004-50589 Supplier Medchemexpress LLC Supplier No. 25MGHYB0318R
Metronidazole is an orally active nitroimidazole antibiotic that can cross the blood-brain barrier. It is used for research on anaerobic infections.
  • Inhibits anaerobic protozoa such as Trichomonas vaginalis, Entamoeba histolytica, Giardia lamblia, and Balantidium coli
  • Inhibits anaerobic bacteria and demonstrates good bactericidal activity
  • Induces granular formation and triggers apoptosis in Blastocystis sp
  • Crosses the blood-brain barrier for neurotoxicity research