Learn More
Cayman Chemical Metronidazole, Purity: ≥98%, CAS: 443-48-1, 5 g
SDP
Catalog No. 502897034
Encompass_Preferred
An antibiotic; has activity against anaerobic bacteria and protozoa; reduces the growth of E. coli in vitro (MIC = 128 mg/L under anaerobic conditions); reduces viable counts of B. fragilis in a rabbit model of infection
Catalog No. 50-289-7034 Supplier Cayman Chemical Supplier No. 90024095 G
An antibiotic; has activity against anaerobic bacteria and protozoa; reduces the growth of E. coli in vitro (MIC = 128 mg/L under anaerobic conditions); reduces viable counts of B. fragilis in a rabbit model of infection