Learn More
TARGETMOL CHEMICALS INC METRONIDAZOLE BENZOATE 100MG
SDP
Catalog No. 502581489
Encompass_Preferred
Also available in and bulk. Please contact Fisher for quotes. Metronidazole Benzoate (Benzoyl metronidazole) is a derivative of Metronidazole and benzoic acid which has antibacterial anti-parasitic and anti-trichomonas functions and is often used to treat infections caused by anaerobic bacteria. Metronidazole Benzoate is a salt form of Metronidazole an antibiotic compound commonly used to treat bacterial and parasitic infections. purity: 99%
Catalog No. 50-258-1489 Supplier TARGETMOL CHEMICALS INC Supplier No. T12022100MG
Also available in and bulk. Please contact Fisher for quotes. Metronidazole Benzoate (Benzoyl metronidazole) is a derivative of Metronidazole and benzoic acid which has antibacterial anti-parasitic and anti-trichomonas functions and is often used to treat infections caused by anaerobic bacteria. Metronidazole Benzoate is a salt form of Metronidazole an antibiotic compound commonly used to treat bacterial and parasitic infections. purity: 99%