Learn More
Cayman Chemical Metronidazole Benzoate, Purity: ≥98%, CAS: 13182-89-3, 250 mg
SDP
Catalog No. 502948090
Encompass_Preferred
An antibiotic and antiprotozoal agent; active against P. asaccharolyticus, C. perfringens, C. septicum, B. fragilis, and B. thetaiotaomicron (MICs = 3.6, 7.3, 1.8, 7.3, and 7.3 µM, respectively); reduces parasitemia in rats when cultured with T. congolense or T. brucei prior to infection
Catalog No. 50-294-8090 Supplier Cayman Chemical Supplier No. 33316250 MG
An antibiotic and antiprotozoal agent; active against P. asaccharolyticus, C. perfringens, C. septicum, B. fragilis, and B. thetaiotaomicron (MICs = 3.6, 7.3, 1.8, 7.3, and 7.3 µM, respectively); reduces parasitemia in rats when cultured with T. congolense or T. brucei prior to infection