Learn More
Medchemexpress LLC Metronidazole-d3 | 83413-09-6 | 99.97% | 5 MG
SDP
Catalog No. 5000406589
Encompass_Preferred
Metronidazole-d3 is deuterium labeled metronidazole, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06589 Supplier Medchemexpress LLC Supplier No. HYB0318S25MG
Metronidazole-d3 is deuterium labeled metronidazole, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS