Learn More
Medchemexpress LLC Metronidazole-d3 | 83413-09-6 | MFCD27702816 | 100.0% | 1 MG
SDP
Catalog No. 5000406344
Encompass_Preferred
Metronidazole-d3 is a deuterium-labeled form of Metronidazole. It can be used for research purposes, including as a tracer or an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS. This compound typically appears as a light yellow to yellow solid.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes incorporated into drug molecules.
  • Deuteration offers potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06344 Supplier Medchemexpress LLC Supplier No. HYB0318S21MG
Metronidazole-d3 is a deuterium-labeled form of Metronidazole. It can be used for research purposes, including as a tracer or an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS. This compound typically appears as a light yellow to yellow solid.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes incorporated into drug molecules.
  • Deuteration offers potential to affect pharmacokinetic and metabolic profiles of drugs.