Learn More
Medchemexpress LLC Metronidazole-d4 | 1261392-47-5 | 98.3% | 1 MG
SDP
Catalog No. 5000406883
Encompass_Preferred
Metronidazole-d4 is a deuterium-labeled Metronidazole, a nitroimidazole antibiotic medication used for anaerobic bacteria and protozoa. It is for research use only and not sold to patients. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06883 Supplier Medchemexpress LLC Supplier No. HYB0318S11MG
Metronidazole-d4 is a deuterium-labeled Metronidazole, a nitroimidazole antibiotic medication used for anaerobic bacteria and protozoa. It is for research use only and not sold to patients. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.