Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metronidazole impurity 1 | 5006-68-8 | MFCD00159670 | 98.0% | 157.13 | C5H7N3O3 | 100 MG
SDP

Catalog No. 5000302562
Encompass_Preferred

Metronidazole impurity 1 is an impurity of metronidazole supplied for research and analytical applications. It is intended for use as an analytical reference or standard in impurity profiling, method development, and quality-control testing of metronidazole products.

  • Used as an analytical reference for metronidazole impurity profiling.
  • Chemical formula C5H7N3O3.
  • Molecular weight 157.13 g/mol.
  • CAS number 5006-68-8.
  • Purity typically reported as 98.0% by HPLC.
  • Store under conditions specified on the certificate of analysis.

Catalog No. 50-003-02562 Supplier Medchemexpress LLC Supplier No. HYW015509100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metronidazole impurity 1 is an impurity of metronidazole supplied for research and analytical applications. It is intended for use as an analytical reference or standard in impurity profiling, method development, and quality-control testing of metronidazole products.

  • Used as an analytical reference for metronidazole impurity profiling.
  • Chemical formula C5H7N3O3.
  • Molecular weight 157.13 g/mol.
  • CAS number 5006-68-8.
  • Purity typically reported as 98.0% by HPLC.
  • Store under conditions specified on the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.