Learn More
Cayman Chemical Metronidazole Impurity G, Purity: ≥95%, CAS: 1010-93-1, 10 mg
SDP
Catalog No. 502943483
Encompass_Preferred
A potential impurity found in commercial preparations of metronidazole; an inactive metabolite of metronidazole
Catalog No. 50-294-3483 Supplier Cayman Chemical Supplier No. 4229110 MG
A potential impurity found in commercial preparations of metronidazole; an inactive metabolite of metronidazole