Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metronidazole, MilliporeSigma™ Supelco™

Catalog No. 111012203 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2203 5 g
11-101-2201 25 g
11-101-2202 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-2203 Supplier MilliporeSigma Supelco Supplier No. M37615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical structure: imidazole

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 443-48-1
Physical Form Crystalline
Linear Formula C6H9N3O3
MDL Number MFCD00009750
Melting Point 159°C to 161°C
Synonym 2-Methyl-5-nitroimidazole-1-ethanol
Type Analytical Standard
Quantity 5 g
Formula Weight 171.15
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.