Learn More
Medchemexpress LLC Metronidazole (Standard) | 443-48-1 | MFCD00009750 | 99.99% | 50 MG
SDP
Catalog No. 5000406893
Encompass_Preferred
Metronidazole (Standard) is an orally active nitroimidazole antibiotic and an analytical standard intended for research and analytical applications. It can cross the blood-brain barrier and is used for research on anaerobic infections.
  • Analytical standard for Metronidazole
  • Suitable for research and analytical applications
  • Used as a reference standard in assays
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS studies
  • Orally active nitroimidazole antibiotic
  • Can cross the blood-brain barrier
  • Used for research on anaerobic infections
Catalog No. 50-004-06893 Supplier Medchemexpress LLC Supplier No. HYB0318R50MG
Metronidazole (Standard) is an orally active nitroimidazole antibiotic and an analytical standard intended for research and analytical applications. It can cross the blood-brain barrier and is used for research on anaerobic infections.
  • Analytical standard for Metronidazole
  • Suitable for research and analytical applications
  • Used as a reference standard in assays
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS studies
  • Orally active nitroimidazole antibiotic
  • Can cross the blood-brain barrier
  • Used for research on anaerobic infections