Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metronidazole USP, Medisca™
GREENER_CHOICESDP

Catalog No. 18048131 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
25 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-131 1 kg
18-048-024 5 g
18-048-027 500 g
18-048-064 25 kg
18-048-025 25 g
18-048-026 100 g
18-048-083 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-048-131 Supplier Medisca Supplier No. 014609
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metronidazole USP can be white to pale yellow crystals or crystalline powder. It is odorless and is stable in air, but darkens on exposure to light.

Chemical Identifiers

CAS 443-48-1

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White to Light Yellow
Quantity 1 kg
Percent Purity 99.0 to 101.0%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.