Learn More
Cayman Chemical Metronidazoled4 GC MS Standard
SDP
Catalog No. 502888546
Encompass_Preferred
An internal standard for the quantification of metronidazole by GC- or LC-MS
Catalog No. 50-288-8546 Supplier Cayman Chemical Supplier No. 100106591 MG
An internal standard for the quantification of metronidazole by GC- or LC-MS