Learn More
Med Vet International Blood Glucose Meter True Metrix™ 4 Second Results Stores up to 500 Results No Coding Required, RE4H01-40, Each
Catalog No. 503045534
Encompass_Preferred
4 second results 0.5 microliter sample from fingertip or forearm Stores up to 500 results with date and time No coding required
Catalog No. 50-304-5534 Supplier Med Vet International Supplier No. 984036EA
4 second results 0.5 microliter sample from fingertip or forearm Stores up to 500 results with date and time No coding required