Learn More
Selleck Chemical LLC Metroprolol succinate S5341-25mg
SDP
Catalog No. 503110180
Encompass_Preferred
Metroprolol Succinate (Metoprolol succinate) is the succinate salt form of metoprolol a cardioselective competitive beta-1 adrenergic receptor antagonist with antihypertensive properties and devoid of intrinsic sympathomimetic activity
Catalog No. 50-311-0180 Supplier Selleck Chemical LLC Supplier No. S534125MG
Metroprolol Succinate (Metoprolol succinate) is the succinate salt form of metoprolol a cardioselective competitive beta-1 adrenergic receptor antagonist with antihypertensive properties and devoid of intrinsic sympathomimetic activity