Learn More
Metrohm USA METROSEP 4/5 S-GUARD
SDP
Supplier:  Metrohm USA 61006540
Encompass
The Metrosep A Supp 4/5 S-Guard/4.0 reliably protects the Metrosep A Supp 4, 5 and 7 IC anion columns against contamination from the sample or eluent.
Catalog No. NC0661362