Learn More
Metrohm USA Metrosep A PCC2 VHC/4.0 Spec Column Concentrator
SDP
Supplier:  Metrohm USA 6.1006.350
Encompass
For anion preconcentration and matrix elimination. The very high capacity is required primarily when matrix effects might cause an overloading of the preconcentration column or when samples with high ionic strength are to be analyzed.
Catalog No. NC3348150