Learn More
Metrohm USA METROSEP A SUPP 4/5 GUARD
SDP
Catalog No. MP61006500 Shop All Metrohm USA Products
Encompass
The Metrosep a Supp 4/5 Guard/4.0 reliably protects the Metrosep a Supp 4, 5 and 7 IC anion columns against contamination from the sample or the eluent. It contains the same separation material as the Metrosep a Supp 5, is also made of PEEK, and is screwed directly onto the respective separation column with nearly no dead volume ( in.On Column Guard System in.). The guard column prolongs the service life of the analytical column, with practically no influence on its chromatographic separating efficiency. The economical price and simple handling make using the a Supp 4/5 Guard/4.0 highly recommended.
Catalog No. MP61006500 Supplier Metrohm USA Supplier No. 61006500
You must be signed in and have a business account to purchase online.
The Metrosep a Supp 4/5 Guard/4.0 reliably protects the Metrosep a Supp 4, 5 and 7 IC anion columns against contamination from the sample or the eluent. It contains the same separation material as the Metrosep a Supp 5, is also made of PEEK, and is screwed directly onto the respective separation column with nearly no dead volume ( in.On Column Guard System in.). The guard column prolongs the service life of the analytical column, with practically no influence on its chromatographic separating efficiency. The economical price and simple handling make using the a Supp 4/5 Guard/4.0 highly recommended.