Learn More
Metrohm USA METROSEP A SUPP 5 - 100
SDP
Supplier:  Metrohm USA 61006510
Encompass
The Metrosep A Supp 5 - 100/4.0 allows highly efficient, rapid separations. This property makes the Metrosep A Supp 5 - 100/4.0 the standard column for short analysis times and the determination of anions which elute late (e.g., perchlorate).
Catalog No. MP61006510