Learn More
Metrohm USA METROSEP C 4 S GUARD
SDP
Supplier:  Metrohm USA 61050420
Encompass
The Metrosep C 4 - 150/4.0 is the universal standard column in cation analysis for accomplishing high separating efficiency in a short time. The Metrosep C 4 - 150/4.0 is the ideal separation column for the analysis of alkaline and earth alkaline metals in aqueous media.
Catalog No. NC9979639