Learn More
Fine Science Tools Metzenbaum Baby Scissors, Stainless Steel, 13 cm
SDP
Catalog No. NC9822043
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 31mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Serrated: No
Catalog No. NC9822043 Supplier Fine Science Tools Supplier No. 1401813
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 31mm/ Alloy / Material: Stainless Steel/ Length: 13cm/ Serrated: No