Learn More
Medchemexpress LLC Metsulfuron-methyl | 74223-64-6 | 99.6% | 100 MG
SDP
Catalog No. 5000410079
Encompass_Preferred
Metsulfuron-methyl is a systemic sulfonylurea herbicide widely utilized for controlling broadleaf weeds and annual grasses in crops such as rice, corn, wheat, and barley.
  • Demonstrates high herbicidal activity
  • Low mammalian toxicity, with an LD50 for rats exceeding 5000 mg/kg
Catalog No. 50-004-10079 Supplier Medchemexpress LLC Supplier No. HYB1869100MG
Metsulfuron-methyl is a systemic sulfonylurea herbicide widely utilized for controlling broadleaf weeds and annual grasses in crops such as rice, corn, wheat, and barley.
  • Demonstrates high herbicidal activity
  • Low mammalian toxicity, with an LD50 for rats exceeding 5000 mg/kg