Learn More
Aviva Systems Biology Corp METT5D1 Antibody - N-terminal region (ARP49066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against METT5D1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human METT5D1. Alternate names: FLJ33979, METT5D1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60008